Oxybenzone ≥98%

Supplier: BeanTown Chemical

Synonyms: Eusolex® 4360, 2-Hydroxy-4-methoxybenzophenone, (2-Hydroxy-4-methoxyphenyl)(phenyl)methanone

212975-1KG 212975-250G 212975-50G
BT212975-1KGEA 223 USD
BT212975-1KG BT212975-250G BT212975-50G
Oxybenzone ≥98%
Oxybenzone

CAS: 131-57-7; EC No: 205-031-5; MDL No: MFCD00008387; RTECS: DJ1575000 Crystalline; Molecular Formula: C14H12O3; MW: 228.24 Melting Point: 62-64°; Boiling Point: 150-160°/5 mmHg; Flash point: 100°C (212°F)


  • RTECS: DJ1575000
  • Crystalline

Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
Boiling Pt: 150…160 °C/5 mmHg
Melting Pt: 62…64 °C
Flash Pt: 100 °C (212 °F)
MDL Number: MFCD00008387
CAS Number: 131-57-7
EINECS: 205-031-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR