D-(+)-Menthol ≥99%

Supplier: BeanTown Chemical

Synonyms: (1S)-(+)-Menthol, (1R,2S,4S)-(+)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, (1S,2R,5S)-(+)-Menthol, D(+)-Menthyl alcoholol

212905-100G 212905-25G
BT212905-100GEA 164 USD
BT212905-100G BT212905-25G
D-(+)-Menthol ≥99%
D-(+)-Menthol

CAS: 15356-60-2; EC No: 239-387-8; MDL No: MFCD00062983; RTECS: OT0350000 Crystalline; Molecular Formula: C10H20O; MW: 156.27 Melting Point: 43-44°; Boiling Point: 103-104°/9 mmHg; Flash point: 101°C (214°F) Optical Rotation: [α]23/D +48°, c = 10 in ethanol


  • RTECS: OT0350000
  • Optical rotation: [α]23/D +48°, c = 10 in ethanol
  • Crystalline

Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 103…104 °C/9 mmHg
Melting Pt: 43…44 °C
Flash Pt: 101 °C (214 °F)
MDL Number: MFCD00062983
CAS Number: 15356-60-2
EINECS: 239-387-8

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR