1,3,5-Tribromobenzene ≥98%
Supplier: BeanTown Chemical
212880-100G
212880-25G
212880-500G
BT212880-100GEA
136
USD
BT212880-100G
BT212880-25G
BT212880-500G
1,3,5-Tribromobenzene ≥98%
1,3,5-Tribromobenzene
CAS: 626-39-1; EC No: 210-947-3; MDL No: MFCD00000080; RTECS: DC1957200
Powder; Molecular Formula: C6H3Br3; MW: 314.80
Melting Point: 117-121°; Boiling Point: 271°
- RTECS: DC1957200
- Powder
Formula:
C₆H₃Br₃ Boiling Pt: 271 °C Melting Pt: 117…121 °C |
MDL Number:
MFCD00000080 CAS Number: 626-39-1 EINECS: 210-947-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...