2-Bromo-1,3-dimethoxybenzene ≥98%

Supplier: BeanTown Chemical

212855-1G 212855-25G 212855-5G
BT212855-1GEA 48 USD
BT212855-1G BT212855-25G BT212855-5G
2-Bromo-1,3-dimethoxybenzene ≥98%
2-Bromo-1,3-dimethoxybenzene

CAS: 16932-45-9; MDL No: MFCD07780177 Solid; Molecular Formula: C8H9BrO2; MW: 217.06 Melting Point: 92-94°


  • Solid

Formula: C₈H₉BrO₂
MW: 217.06 g/mol
Melting Pt: 92…94 °C
MDL Number: MFCD07780177
CAS Number: 16932-45-9

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR