Triphenylmethylamine ≥98%

Supplier: BeanTown Chemical

Synonyms: Triphenylaminomethane, Tritylamine, Triphenylmethanamine

212835-1G 212835-25G 212835-5G
BT212835-1GEA 26 USD
BT212835-1G BT212835-25G BT212835-5G
Triphenylmethylamine ≥98%
Triphenylmethylamine

CAS: 5824-40-8; MDL No: MFCD00008047 Crystalline; Linear Formula: (C6H5)3CNH2; Molecular Formula: C19H17N; MW: 259.35 Melting Point: 102-104°; Boiling Point: 223°/14 mmHg Air Sensitive


  • Air Sensitive
  • Crystalline

Formula: C₁₉H₁₇N
MW: 259.35 g/mol
Boiling Pt: 223 °C/14 mmHg
Melting Pt: 102…104 °C
MDL Number: MFCD00008047
CAS Number: 5824-40-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR