Strontium chloride hexahydrate ≥99%
Supplier: BeanTown Chemical
212815-1KG
212815-250G
BT212815-1KGEA
215.4
USD
BT212815-1KG
BT212815-250G
Strontium chloride hexahydrate ≥99%
Strontium chloride hexahydrate
CAS: 10025-70-4; EC No: 233-971-6; MDL No: MFCD00149865; RTECS: WK8575000
Crystalline; Linear Formula: SrCl2·6H2O ; MW: 266.62
Density (g/mL): 1.93; Refractive Index: 1.650
Hygroscopic
- RTECS: WK8575000
- Refractive index: 1.650
- Hygroscopic
- Crystalline
Formula:
SrCl₂·6H₂O Density: 1.93 g/mL |
MDL Number:
MFCD00149865 CAS Number: 10025-70-4 EINECS: 233-971-6 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...