(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane ≥98%

Supplier: BeanTown Chemical

Synonyms: tert-butyl (1S,4S)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate

212785-1G 212785-250MG 212785-5G
BT212785-1GEA 142 USD
BT212785-1G BT212785-250MG BT212785-5G
(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane ≥98%
(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane

CAS: 113451-59-5; MDL No: MFCD01569250 Powder; Molecular Formula: C10H18N2O2; MW: 198.26 Melting Point: 74-76° Optical Rotation: [α]22/D -44°, c = 1 in chloroform Air Sensitive


  • Optical rotation: [α]22/D -44°, c = 1 in chloroform
  • Air Sensitive
  • Powder

Formula: C₁₀H₁₈N₂O₂
MW: 198.26 g/mol
Melting Pt: 74…76 °C
MDL Number: MFCD01569250
CAS Number: 113451-59-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR