Benzoic acid ≥99%
Supplier: BeanTown Chemical
212775-1KG
212775-250G
212775-5KG
BT212775-1KGEA
46.39
USD
BT212775-1KG
BT212775-250G
BT212775-5KG
Benzoic acid ≥99%
Benzoic acid
CAS: 65-85-0; EC No: 200-618-2; MDL No: MFCD00002398; RTECS: DG0875000
Crystalline; Linear Formula: C6H5CO2H; MW: 122.12
Melting Point: 121-125°; Boiling Point: 249°; Flash point: 121°C (250°F)
- RTECS: DG0875000
- Crystalline
Formula:
C₇H₆O₂ Boiling Pt: 249 °C Melting Pt: 121…125 °C Flash Pt: 121 °C (250 °F) |
MDL Number:
MFCD00002398 CAS Number: 65-85-0 EINECS: 200-618-2 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...