(Pentamethylcyclopentadienyl)rhodium(III) dichloride dimer ≥99%

Supplier: BeanTown Chemical

Synonyms: Dichloro(pentamethylcyclopentadienyl)rhodium (III) dimer, [Rh(Cp-)Cl2]2, Dichloro(pentamethylcyclopentadienyl)rhodium(III) dimer, Dichloro(pentamethylcyclopentadienyl)rhodium(III) Dimer

212770-1G 212770-250MG
BT212770-1GEA 649 USD
BT212770-1G BT212770-250MG
(Pentamethylcyclopentadienyl)rhodium(III) dichloride dimer ≥99%
(Pentamethylcyclopentadienyl)rhodium(III) dichloride dimer

CAS: 12354-85-7; MDL No: MFCD00061552 Powder; Molecular Formula: C10H15Cl4Rh2 ; MW: 618.08


  • Powder

Formula: C₂₀H₃₀Cl₄Rh₂
MW: 618.07 g/mol
MDL Number: MFCD00061552
CAS Number: 12354-85-7

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR