4-Aminoantipyrine ≥97%

Supplier: BeanTown Chemical

Synonyms: 4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Ampyrone, 4-Aminophenazone

212760-100G 212760-500G
BT212760-100GEA 60.88 USD
BT212760-100G BT212760-500G
4-Aminoantipyrine ≥97%
4-Aminoantipyrine

CAS: 83-07-8; EC No: 201-452-3; MDL No: MFCD00003145; RTECS: CD2480000 Solid; Molecular Formula: C11H13N3O; MW: 203.24 Melting Point: 105-110°


  • RTECS: CD2480000
  • Solid

Formula: C₁₁H₁₃N₃O
MW: 203.24 g/mol
Melting Pt: 105…110 °C
MDL Number: MFCD00003145
CAS Number: 83-07-8
EINECS: 201-452-3

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR