(2,4-Dichlorophenyl)acetic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: DCAA, 2,4-Dichlorophenylacetic acid

212735-100G 212735-25G 212735-5G
BT212735-100GEA 171 USD
BT212735-100G BT212735-25G BT212735-5G
(2,4-Dichlorophenyl)acetic acid ≥98%
(2,4-Dichlorophenyl)acetic acid

CAS: 19719-28-9; EC No: 243-248-7; MDL No: MFCD00004318 Crystalline; Molecular Formula: C8H6Cl2O2; MW: 205.04 Melting Point: 129-131°


  • Crystalline

Formula: (C₆H₃Cl₂)CH₂COOH
MW: 205.04 g/mol
Melting Pt: 129…131 °C
MDL Number: MFCD00004318
CAS Number: 19719-28-9
EINECS: 243-248-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR