4-(Boc-amino)-2-chloropyridine ≥95%
Supplier: BeanTown Chemical
Synonyms:
2-Chloro-4-(tert-butoxycarbonyl)pyridine, tert-Butyl (2-chloropyridin-4-yl)carbamate, 4-Amino-2-chloropyridine, 4-BOC protected, tert-Butyl 2-chloropyridine-4-carbamate
212730-1G
212730-5G
BT212730-1GEA
205
USD
BT212730-1G
BT212730-5G
4-(Boc-amino)-2-chloropyridine ≥95%
4-(Boc-amino)-2-chloropyridine
CAS: 234108-73-7; MDL No: MFCD02179232
Crystalline/Powder; Molecular Formula: C10H13ClN2O2 ; MW: 228.68
- Crystalline/Powder
Formula:
C₁₀H₁₃ClN₂O₂ MW: 228.68 |
MDL Number:
MFCD02179232 CAS Number: 234108-73-7 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...