Tetraethylammonium fluoride hydrate ≥97%
Supplier: BeanTown Chemical
Synonyms:
Tetrylammonium fluoride hydrate, Tetraethylammonium fluoride xhydrate
212705-100G
212705-25G
212705-5G
BT212705-100GEA
388.16
USD
BT212705-100G
BT212705-25G
BT212705-5G
Tetraethylammonium fluoride hydrate ≥97%
Tetraethylammonium fluoride hydrate
CAS: 98330-04-2; EC No: 211-559-7; MDL No: MFCD00149990
Crystalline; Linear Formula: (CH3CH2)4N(F)·xH2O; MW: 149.26 (anhydrous)
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
C₈H₂₀FN·xH₂O MW: 149.25 g/mol (anhy) |
MDL Number:
MFCD00149990 CAS Number: 98330-04-2 EINECS: 211-559-7 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...