Ethyl hydrogen malonate ≥90%, Technical Grade
Supplier: BeanTown Chemical
212700-100G
212700-25G
212700-5G
BT212700-100GEA
334.62
USD
BT212700-100G
BT212700-25G
BT212700-5G
Ethyl hydrogen malonate ≥90%, Technical Grade
Ethyl hydrogen malonate
CAS: 1071-46-1; EC No: 213-992-7; MDL No: MFCD00020490
Liquid; Linear Formula: HO2CCH2CO2CH2CH3; Molecular Formula: C5H8O4; MW: 132.12
Boiling Point: 106°/3 mmHg; Flash point:
- Refractive index: 1.435
- Liquid
Formula:
C₅H₈O₄ Boiling Pt: 106 °C/3 mmHg Density: 1.119 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00020490 CAS Number: 1071-46-1 EINECS: 213-992-7 |
Specification Test Results
Assay | 90% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...