2-(Boc-amino)pyridine-3-boronic acid pinacol ester ≥98%
Supplier: BeanTown Chemical
212695-1G
212695-250MG
BT212695-1GEA
256
USD
BT212695-1G
BT212695-250MG
2-(Boc-amino)pyridine-3-boronic acid pinacol ester ≥98%
2-(Boc-amino)pyridine-3-boronic acid pinacol ester
CAS: 1072944-99-0; MDL No: MFCD08063072
Powder; Molecular Formula: C16H25BN2O4; MW: 320.19
Air Sensitive
- Air Sensitive
- Powder
Formula:
C₁₆H₂₅BN₂O₄ |
MDL Number:
MFCD08063072 CAS Number: 1072944-99-0 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...