3,5-Dibenzyloxybenzoic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 3,5-Bis(benzyloxy)benzoic acid

212650-25G 212650-5G
BT212650-25GEA 324 USD
BT212650-25G BT212650-5G
3,5-Dibenzyloxybenzoic acid ≥98%
3,5-Dibenzyloxybenzoic acid

CAS: 28917-43-3; EC No: 249-311-5; MDL No: MFCD00191341 Solid; Molecular Formula: C21H18O4; MW: 334.37 Boiling Point: 470°


  • Solid

Formula: C₂₁H₁₈O₄
MW: 334.37 g/mol
Boiling Pt: 470 °C
MDL Number: MFCD00191341
CAS Number: 28917-43-3
EINECS: 249-311-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR