3,5-Di-tert-butyl-4-hydroxybenzoic acid ≥98%
Supplier: BeanTown Chemical
212645-10G
212645-50G
BT212645-10GEA
37
USD
BT212645-10G
BT212645-50G
3,5-Di-tert-butyl-4-hydroxybenzoic acid ≥98%
3,5-Di-tert-butyl-4-hydroxybenzoic acid
CAS: 1421-49-4; EC No: 215-823-2; MDL No: MFCD00008827; RTECS: DG6320000
Crystalline/Powder; Molecular Formula: C15H21O3; MW: 250.33
Melting Point: 206-209°
- RTECS: DG6320000
- Crystalline/Powder
Formula:
C₁₅H₂₂O₃ MW: 250.34 g/mol Melting Pt: 206…209 °C |
MDL Number:
MFCD00008827 CAS Number: 1421-49-4 EINECS: 215-823-2 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...