4-Bromo-2-chloropyridine ≥94%

Supplier: BeanTown Chemical

212635-25G 212635-5G
BT212635-25GEA 292 USD
BT212635-25G BT212635-5G
4-Bromo-2-chloropyridine ≥94%
4-Bromo-2-chloropyridine

CAS: 73583-37-6; MDL No: MFCD03840756 Crystalline; Molecular Formula: C5H3BrClN ; MW: 192.44 Flash point: 107°C (225°F) Density (g/mL): 1.734; Refractive Index: 1.590


  • Refractive index: 1.590
  • Crystalline

Formula: C₅H₃BrClN
MW: 192.44 g/mol
Density: 1.734 g/mL
Flash Pt: 107 °C (225 °F)
MDL Number: MFCD03840756
CAS Number: 73583-37-6

Order Now

Specification Test Results

Assay 94%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR