Indium(III) sulfide ≥99.95% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Diindium trisulphide, Indium trisulphide

212590-1G 212590-25G 212590-5G
BT212590-1GEA 18 USD
BT212590-1G BT212590-25G BT212590-5G
Indium(III) sulfide ≥99.95% (trace metals basis)
Indium(III) sulphide

CAS: 12030-24-9; EC No: 234-742-3; MDL No: MFCD00011061 Powder; Molecular Formula: In2S3; MW: 325.83 Moisture Sensitive


  • Moisture Sensitive
  • Powder

Formula: In₂S₃
MW: 325.84 g/mol
MDL Number: MFCD00011061
CAS Number: 12030-24-9
EINECS: 234-742-3

Order Now

Specification Test Results

Assay 99.95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR