(R)-(+)-2-Amino-3-methyl-1,1-diphenyl-1-butanol ≥95%
Supplier: BeanTown Chemical
212545-1G
212545-5G
BT212545-1GEA
135
USD
BT212545-1G
BT212545-5G
(R)-(+)-2-Amino-3-methyl-1,1-diphenyl-1-butanol ≥95%
(R)-(+)-2-Amino-3-methyl-1,1-diphenyl-1-butanol
CAS: 86695-06-9; MDL No: MFCD01318247
Powder; Linear Formula: (CH3)2CHCH(NH2)C(C6H5)2(OH); Molecular Formula: C17H21NO; MW: 255.35
Melting Point: 95-99°
Optical Rotation: [α]20/D +132°, c = 1 in chloroform
- Optical rotation: [α]20/D +132°, c = 1 in chloroform
- Powder
Formula:
C₁₇H₂₁NO MW: 255.3600 g/mol Melting Pt: 95…99 °C |
MDL Number:
MFCD01318247 CAS Number: 86695-06-9 |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...