2-Fluoro-4-(trans-4-pentylcyclohexyl)-4'-(trans-4-propylcyclohexyl)biphenyl ≥97%
Supplier: BeanTown Chemical
212535-1G
212535-5G
BT212535-1GEA
103
USD
BT212535-1G
BT212535-5G
2-Fluoro-4-(trans-4-pentylcyclohexyl)-4'-(trans-4-propylcyclohexyl)biphenyl ≥97%
2-Fluoro-4-(trans-4-pentylcyclohexyl)-4'-(trans-4-propylcyclohexyl)biphenyl
CAS: 106349-49-9; MDL No: MFCD11053473
Solid; Molecular Formula: C32H45F; MW: 448.71
Melting Point: 79-81°
- Solid
Formula:
C₃₂H₄₅F Melting Pt: 79…81 °C |
MDL Number:
MFCD11053473 CAS Number: 106349-49-9 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...