2,4-Dichloro-3-methyl benzoic acid ≥99%
Supplier: BeanTown Chemical
Synonyms:
2,4-Dichloro-3-methylbenzoic acid
212530-1G
212530-5G
BT212530-1GEA
318
USD
BT212530-1G
BT212530-5G
2,4-Dichloro-3-methyl benzoic acid ≥99%
2,4-Dichloro-3-methyl benzoic acid
CAS: 83277-23-0
Solid; Molecular Formula: C8H6Cl2O2; MW: 205.04
- Solid
Formula:
C₈H₆Cl₂O₂ MW: 205.034 g/mol |
MDL Number:
MFCD16659660 CAS Number: 83277-23-0 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...