1,13-Tridecanedioic acid ≥94%

Supplier: BeanTown Chemical

Synonyms: Tridecanedioic acid, Undecane-1,11-dicarboxylic acid, 1,11-Undecanedicarboxylic acid

212510-1G 212510-5G
BT212510-1GEA 17 USD
BT212510-1G BT212510-5G
1,13-Tridecanedioic acid ≥94%
1,13-Tridecanedioic acid

CAS: 505-52-2; EC No: 208-011-4; MDL No: MFCD00002740 Powder; Linear Formula: HO2C(CH2)11CO2H ; Molecular Formula: C13H24O4; MW: 244.33 Melting Point: 112-114°


  • Powder

Formula: C₁₃H₂₄O₄
MW: 244.33 g/mol
Melting Pt: 112…114 °C
MDL Number: MFCD00002740
CAS Number: 505-52-2
EINECS: 208-011-4

Order Now

Specification Test Results

Assay 94%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR