5-Methyl-2-(trifluoromethyl)benzoic acid ≥97%
Supplier: BeanTown Chemical
212445-1G
212445-5G
BT212445-1GEA
141
USD
BT212445-1G
BT212445-5G
5-Methyl-2-(trifluoromethyl)benzoic acid ≥97%
5-Methyl-2-(trifluoromethyl)benzoic acid
CAS: 120985-68-4; MDL No: MFCD06660291
Powder; Molecular Formula: C9H7F3O2; MW: 204.15
Melting Point: 90-94°
- Powder
Formula:
C₉H₇F₃O₂ Melting Pt: 90…94 °C |
MDL Number:
MFCD06660291 CAS Number: 120985-68-4 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...