Cesium chloride 99,9

Supplier: BeanTown Chemical

Synonyms: Cesium Chloride

212380-100G 212380-25G
BT212380-100GEA 194.72 USD
BT212380-100G BT212380-25G
Cesium chloride 99,9
Cesium chloride

CAS: 7647-17-8; EC No: 231-600-2; MDL No: MFCD00010955; RTECS: FK9625000 Crystalline; Linear Formula: CsCl; MW: 168.36 Melting Point: 645° Hygroscopic


  • RTECS: FK9625000
  • Hygroscopic
  • Crystalline

Formula: CsCl
MW: 168.36 g/mol
Boiling Pt: 1303
Melting Pt: 646
MDL Number: MFCD00010955
CAS Number: 7647-17-8
EINECS: 231-600-2
Merck Index: 13,02021

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR