1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide ≥98%

Supplier: BeanTown Chemical

212350-1G 212350-25G 212350-5G
BT212350-1GEA 26 USD
BT212350-1G BT212350-25G BT212350-5G
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide ≥98%
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide

CAS: 22572-40-3; EC No: 245-095-1; MDL No: MFCD00011833 Solid; Linear Formula: CH3CH2N=C=N(CH2)3N(CH3)3I; Molecular Formula: C9H20IN3; MW: 297.18 Melting Point: 97-99° Hygroscopic , Light Sensitive


  • Hygroscopic, Light Sensitive
  • Solid

Formula: CH₃CH₂N=C=N(CH₂)₃N(CH₃)₃I
Melting Pt: 97…99 °C
MDL Number: MFCD00011833
CAS Number: 22572-40-3
EINECS: 245-095-1

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR