Disodium malonate monohydrate

Supplier: BeanTown Chemical

Synonyms: Sodium malonate dibasic monohydrate, Malonic acid disodium salt hydrate

212340-100G 212340-25G
BT212340-100GEA 103 USD
BT212340-100G BT212340-25G
Disodium malonate monohydrate
Disodium malonate monohydrate

CAS: 26522-85-0; EC No: 205-514-0; MDL No: MFCD00002708 Crystalline; Linear Formula: CH2(COONa)2·H2O; MW: 166.04 Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: C₃H₂Na₂O₄·1H₂O
MDL Number: MFCD00149114
CAS Number: 26522-85-0
EINECS: 205-514-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR