Platinum iridium alloy (80:20 w%) ≥99.9% (trace metals basis), wire, annealed, Diameter 0.2 mm

Supplier: BeanTown Chemical

212335-10CM 212335-50CM
BT212335-10CMEA 65.52 USD
BT212335-10CM BT212335-50CM
Platinum iridium alloy (80:20 w%) ≥99.9% (trace metals basis), wire, annealed, Diameter 0.2 mm
Platinum iridium alloy (80:20 w%)

MDL No: MFCD02091683 Solid; Formula: Pt:Ir; 80:20 wt%


  • Solid

MDL Number: MFCD02091683
UN: 3178
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR