1,2-Diiodobenzene ≥98%

Supplier: BeanTown Chemical

212315-1G 212315-25G 212315-5G
BT212315-1GEA 35 USD
BT212315-1G BT212315-25G BT212315-5G
1,2-Diiodobenzene ≥98%
1,2-Diiodobenzene

CAS: 615-42-9; EC No: 210-425-5; MDL No: MFCD00001038 Liquid; Molecular Formula: C6H4I2; MW: 329.90 Boiling Point: 152°/15 mmHg; Flash point: 110°C (230°F) Density (g/mL): 2.524; Refractive Index: 1.718 Light Sensitive


  • Refractive index: 1.718
  • Light Sensitive
  • Liquid

Formula: C₆H₄I₂
MW: 329.91 g/mol
Boiling Pt: 285
Density: 2.524 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00001038
CAS Number: 615-42-9
EINECS: 210-425-5

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR