Chloro(1,5-cyclooctadiene)rhodium(I) dimer

Supplier: BeanTown Chemical

Synonyms: (1,5-Cyclooctadiene)rhodium chloride dimer

212300-1G 212300-250MG
BT212300-1GEA 1318 USD
BT212300-1G BT212300-250MG
Chloro(1,5-cyclooctadiene)rhodium(I) dimer
Chloro(1,5-cyclooctadiene)rhodium (I) dimer

CAS: 12092-47-6; EC No: 235-157-6; MDL No: MFCD00012415 Crystalline; Molecular Formula: C16H24Cl2Rh2; MW: 493.08 Melting Point: 243° (decomposes) Air Sensitive, Hygroscopic


  • Air Sensitive, Hygroscopic
  • Crystalline

Formula: C₁₆H₂₄Cl₂Rh₂
MW: 493.08 g/mol
Melting Pt: 243 °C (dec.)
MDL Number: MFCD00012415
CAS Number: 12092-47-6
EINECS: 235-157-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR