Tetraethylammonium iodide ≥98%

Supplier: BeanTown Chemical

Synonyms: TEAI, Tetrylammonium iodide

212285-1KG 212285-250G 212285-50G
BT212285-1KGEA 448 USD
BT212285-1KG BT212285-250G BT212285-50G
Tetraethylammonium iodide ≥98%
Tetraethylammonium iodide

CAS: 68-05-3; EC No: 200-676-9; MDL No: MFCD00011829; RTECS: BS7365000 Crystalline; Linear Formula: (CH3CH2)4N(I); Molecular Formula: C8H20IN; MW: 257.17 Melting Point: 300° Light Sensitive, Hygroscopic, Moisture Sensitive


  • RTECS: BS7365000
  • Light Sensitive, Hygroscopic, Moisture Sensitive
  • Crystalline

Formula: C₈H₂₀IN
MW: 257.16 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00011829
CAS Number: 68-05-3
EINECS: 200-676-9

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR