1-Bromo-4-phenylbutane ≥98%

Supplier: BeanTown Chemical

212280-1G 212280-25G 212280-5G
BT212280-1GEA 50 USD
BT212280-1G BT212280-25G BT212280-5G
1-Bromo-4-phenylbutane ≥98%
1-Bromo-4-phenylbutane

CAS: 13633-25-5; MDL No: MFCD00154988 Liquid; Molecular Formula: C10H13Br; MW: 213.11 Density (g/mL): 1.214


  • Liquid

Formula: C₁₀H₁₃Br
Density: 1.214 g/mL
MDL Number: MFCD00154988
CAS Number: 13633-25-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR