2,4-Dichloro-3-nitropyridine ≥95%

Supplier: BeanTown Chemical

212255-1G 212255-5G
BT212255-1GEA 37 USD
BT212255-1G BT212255-5G
2,4-Dichloro-3-nitropyridine ≥95%
2,4-Dichloro-3-nitropyridine

CAS: 5975-12-2 Solid; Molecular Formula: C5H2Cl2N2O2; MW: 192.99


  • Solid

Formula: C₅H₂Cl₂N₂O₂
MW: 192.9876 g/mol
MDL Number: MFCD09261088
CAS Number: 5975-12-2

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR