1,5-Bis(diphenylphosphino)pentane ≥97%

Supplier: BeanTown Chemical

Synonyms: Pentane-1,3-diylbis(diphenylphosphine), Pentylenebis(diphenylphosphine), Bis(1,2-diphenylphosphino)pentane

212225-1G 212225-25G 212225-5G
BT212225-1GEA 40 USD
BT212225-1G BT212225-25G BT212225-5G
1,5-Bis(diphenylphosphino)pentane ≥97%
1,5-Bis(diphenylphosphino)pentane

CAS: 27721-02-4; MDL No: MFCD00003052 Crystalline; Linear Formula: (C6H5)2P(CH2)5P(C6H5)2 ; Molecular Formula: C29H30P2; MW: 440.50 Melting Point: 43-47°; Flash point: 110°C (230°F)


  • Crystalline

Formula: C₂₉H₃₀P₂
MW: 440.5 g/mol
Melting Pt: 43…47 °C
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00003052
CAS Number: 27721-02-4

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR