3-Amino-5-bromopyridine ≥98%
Supplier: BeanTown Chemical
212210-1G
212210-25G
212210-5G
BT212210-1GEA
23
USD
BT212210-1G
BT212210-25G
BT212210-5G
3-Amino-5-bromopyridine ≥98%
3-Amino-5-bromopyridine
CAS: 13535-01-8; MDL No: MFCD01646060
Solid; Molecular Formula: C5H5BrN2 ; MW: 173.01
Melting Point: 65-69°
- Solid
Formula:
C₅H₅BrN₂ Melting Pt: 65…69 °C |
MDL Number:
MFCD01646060 CAS Number: 13535-01-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...