Maleic acid ≥98%

Supplier: BeanTown Chemical

212205-1KG 212205-250G 212205-5KG
BT212205-1KGEA 105.42 USD
BT212205-1KG BT212205-250G BT212205-5KG
Maleic acid ≥98%
Maleic acid

CAS: 110-16-7; EC No: 203-742-5; MDL No: MFCD00063177; RTECS: OM9625000 Powder; Linear Formula: HO2CCH=CHCO2H; Molecular Formula: C4H4O4; MW: 116.07 Melting Point: 132-136°; Boiling Point: 160°; Flash point: 127°C (261°F) Density (g/mL): 1.59


  • RTECS: OM9625000
  • Powder

Formula: C₄H₄O₄
MW: 116.07 g/mol
Boiling Pt: 160 °C
Melting Pt: 132…136 °C
Density: 1.59 g/mL
Flash Pt: 127 °C (261 °F)
MDL Number: MFCD00063177
CAS Number: 110-16-7
EINECS: 203-742-5
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3a1ea1f5bca5d5cf7fd40530f642f47db88908a&vid=1ac7f8cee2cd0433dbfb6e9271d41fe6ecc382c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR