1,1-Dimethylbiguanide hydrochloride ≥97%

Supplier: BeanTown Chemical

Synonyms: AMPK Signaling Activator VII, Metformin, Metformin hydrochloride, 1,1-Dimethylbiguanide Hydrochloride

212165-100G 212165-25G 212165-5G
BT212165-100GEA 261.03 USD
BT212165-100G BT212165-25G BT212165-5G
1,1-Dimethylbiguanide hydrochloride ≥97%
1,1-Dimethylbiguanide hydrochloride

CAS: 1115-70-4; EC No: 214-230-6; MDL No: MFCD00012582; RTECS: DU1800000 Crystalline/Solid; Molecular Formula: C4H12ClN5; MW: 165.62 Melting Point: 223-226°


  • RTECS: DU1800000
  • Crystalline/Solid

Formula: C₄H₁₂ClN₅
MW: 165.625 g/mol
Melting Pt: 223…226 °C
MDL Number: MFCD00012582
CAS Number: 1115-70-4
EINECS: 214-230-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR