Sodium ligninsulfonate, M.W. ∼52000
Supplier: BeanTown Chemical
Synonyms:
Ligninsulphonic acid Sodium Salt
212155-100G
212155-500G
BT212155-100GEA
46.81
USD
BT212155-100G
BT212155-500G
Sodium ligninsulfonate, M.W. ∼52000
Sodium ligninsulfonate
CAS: 8061-51-6; MDL No: MFCD00135572
Solid
- Solid
MDL Number:
MFCD00135572 CAS Number: 8061-51-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...