DL-Valine ≥99%
Supplier: BeanTown Chemical
Synonyms:
H-DL-Val-OH, (±)-Valine, 2-Amino-3-methylbutanoic acid
212145-100G
212145-2.5KG
212145-500G
BT212145-100GEA
38.47
USD
BT212145-100G
BT212145-2.5KG
BT212145-500G
DL-Valine ≥99%
DL-Valine
CAS: 516-06-3; EC No: 208-220-0; MDL No: MFCD00004267
Powder; Molecular Formula: C5H11NO2; MW: 117.15
Melting Point: 295° (decomposes)
- Powder
Formula:
C₅H₁₁NO₂ MW: 117.15 g/mol Boiling Pt: 217 Melting Pt: 86 (dec.) |
MDL Number:
MFCD00004267 CAS Number: 516-06-3 EINECS: 208-220-0 Merck Index: 12,10046 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...