2-Methylpyridin-3-ol ≥99%

Supplier: BeanTown Chemical

Synonyms: 3-Hydroxy-2-methylpyridine

212140-1G 212140-25G 212140-5G
BT212140-1GEA 32 USD
BT212140-1G BT212140-25G BT212140-5G
2-Methylpyridin-3-ol ≥99%
2-Methylpyridin-3-ol

CAS: 1121-25-1; EC No: 214-327-3; MDL No: MFCD00082538 Powder; Molecular Formula: C6H7NO; MW: 109.13 Melting Point: 166-169°


  • Powder

Formula: C₆H₇NO
MW: 109.13 g/mol
Melting Pt: 166…169 °C
MDL Number: MFCD00082538
CAS Number: 1121-25-1
EINECS: 214-327-3

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR