4-Hydroxybenzaldehyde ≥98%

Supplier: BeanTown Chemical

212095-100G 212095-25G 212095-500G
BT212095-100GEA 35 USD
BT212095-100G BT212095-25G BT212095-500G
4-Hydroxybenzaldehyde ≥98%
4-Hydroxybenzaldehyde

CAS: 123-08-0; EC No: 204-599-1; MDL No: MFCD00006939; RTECS: CU6475000 Crystalline; Molecular Formula: C7H6O2; MW: 122.12 Melting Point: 112-116° Air Sensitive


  • RTECS: CU6475000
  • Air Sensitive
  • Crystalline

Formula: C₇H₆O₂
MW: 122.12 g/mol
Melting Pt: 112…116 °C
MDL Number: MFCD00006939
CAS Number: 123-08-0
EINECS: 204-599-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR