11-Bromo-1-undecanol ≥97%
Supplier: BeanTown Chemical
Synonyms:
11-Bromoundecan-1-ol, 11-Bromoundecanol
212070-25G
212070-5G
BT212070-25GEA
90
USD
BT212070-25G
BT212070-5G
11-Bromo-1-undecanol ≥97%
11-Bromo-1-undecanol
CAS: 1611-56-9; EC No: 216-554-3; MDL No: MFCD00004752
Crystalline; Linear Formula: Br(CH2)11OH; Molecular Formula: C11H23BrO; MW: 251.20
Melting Point: 46-49°; Boiling Point: 165-170°/1 mmHg; Flash point:
- Crystalline
Formula:
HO(CH₂)₁₀CH₂Br MW: 251.21 g/mol Boiling Pt: 165…170 °C/1 mmHg Melting Pt: 46…49 °C Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00004752 CAS Number: 1611-56-9 EINECS: 216-554-3 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...