Benzotriazole ≥99%
Supplier: BeanTown Chemical
Synonyms:
1,2,3-Benzotriazole, 1H-1,2,3-Benzotriazole, 1H-Benzotriazole, 1H-Benzo[d][1,2,3]triazole
212060-1KG
212060-250G
212060-50G
BT212060-1KGEA
176
USD
BT212060-1KG
BT212060-250G
BT212060-50G
Benzotriazole ≥99%
Benzotriazole
CAS: 95-14-7; EC No: 202-394-1; MDL No: MFCD00005699; RTECS: DM1225000
Solid; Molecular Formula: C6H5N3; MW: 119.12
Melting Point: 97-99°; Flash point: 170°C (338°F)
- RTECS: DM1225000
- Solid
Formula:
C₆H₅N₃ Melting Pt: 97…99 °C Flash Pt: 170 °C (338 °F) |
MDL Number:
MFCD00005699 CAS Number: 95-14-7 EINECS: 202-394-1 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...