1,4-Oxazepane hydrochloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
Homomorpholine hydrochloride
211990-1G
211990-5G
BT211990-1GEA
274
USD
BT211990-1G
BT211990-5G
1,4-Oxazepane hydrochloride ≥98%
1,4-Oxazepane hydrochloride
CAS: 178312-62-4; MDL No: MFCD02683070
Powder; Molecular Formula: C5H11NO·HCl; MW: 137.61
Melting Point: 158-164°
Hygroscopic
- Hygroscopic
- Powder
Formula:
C₅H₁₂ClNO MW: 137.61 g/mol Melting Pt: 158…164 °C |
MDL Number:
MFCD02683070 CAS Number: 178312-62-4 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...