(±)-α-Lipoic acid ≥98%
Supplier: BeanTown Chemical
211955-25G
211955-5G
BT211955-25GEA
270.37
USD
BT211955-25G
BT211955-5G
(±)-α-Lipoic acid ≥98%
(±)-α-Lipoic acid
CAS: 1077-28-7; EC No: 214-071-2; MDL No: MFCD00005474; RTECS: JP1192000
Solid; Molecular Formula: C8H14O2S2; MW: 206.33
Melting Point: 60-62°
- RTECS: JP1192000
- Solid
Formula:
C₈H₁₄O₂S₂ Melting Pt: 60…62 °C |
MDL Number:
MFCD00005474 CAS Number: 1077-28-7 EINECS: 214-071-2 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...