Diethylene glycol monovinyl ether ≥98%

Supplier: BeanTown Chemical

Synonyms: Vinyl Carbitol, 2-(2-Vinyloxyethoxy)ethanol, Di(ethylene glycol) vinyl ether, 2-(2-(Vinyloxy)ethoxy)ethanol

211940-100ML 211940-10ML
BT211940-100MLEA 293.42 USD
BT211940-100ML BT211940-10ML
Diethylene glycol monovinyl ether ≥98%
Diethylene glycol monovinyl ether

CAS: 929-37-3; MDL No: MFCD00192027; RTECS: KM5495500 Liquid; Linear Formula: H2C=CH(OCH2CH2)2OH; Molecular Formula: C6H12O3; MW: 132.16 Boiling Point: 196°; Flash point: 83°C (181°F) Density (g/mL): 0.968; Refractive Index: 1.448


  • RTECS: KM5495500
  • Refractive index: 1.448
  • Liquid

Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 196 °C
Density: 0.968 g/mL
Flash Pt: 83 °C (181 °F)
MDL Number: MFCD00192027
CAS Number: 929-37-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR