Hafnium dioxide ≥99.99%
Supplier: BeanTown Chemical
211910-1G
211910-5G
BT211910-1GEA
132
USD
BT211910-1G
BT211910-5G
Hafnium dioxide ≥99.99%
Hafnium dioxide
CAS: 12055-23-1; EC No: 235-013-2; MDL No: MFCD00003565
Powder; Molecular Formula: HfO2; MW: 210.49
Density (g/mL): 9.68
- Powder
Formula:
HfO₂ MW: 210.49 Boiling Pt: 5400 Melting Pt: 2812 Density: 9.68 g/mL |
MDL Number:
MFCD00003565 CAS Number: 12055-23-1 EINECS: 235-013-2 Merck Index: 14,04727 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...