2-Aminopyrazine ≥99%

Supplier: BeanTown Chemical

Synonyms: Aminopyrazine, Pyrazin-2-amine

211905-25G 211905-50G 211905-5G
BT211905-25GEA 167 USD
BT211905-25G BT211905-50G BT211905-5G
2-Aminopyrazine ≥99%
2-Aminopyrazine

CAS: 5049-61-6; EC No: 225-748-7; MDL No: MFCD00006137 Crystalline; Molecular Formula: C4H5N3; MW: 95.10 Melting Point: 118-120°


  • Crystalline

Formula: C₄H₅N₃
MW: 95.1 g/mol
Melting Pt: 118…120 °C
MDL Number: MFCD00006137
CAS Number: 5049-61-6
EINECS: 225-748-7

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR