2-Hydroxyethyl methacrylate ≥97% stabilised

Supplier: BeanTown Chemical

Synonyms: HEMA, GMA, Methacrylic acid 2-hydroxyethyl ester, 1,2-Ethanediol mono(2-methylpropenoate), Glycol methacrylate

211890-100G 211890-500G
BT211890-100GEA 26.93 USD
BT211890-100G BT211890-500G
2-Hydroxyethyl methacrylate ≥97% stabilised
2-Hydroxyethyl methacrylate

CAS: 868-77-9; EC No: 212-782-2; MDL No: MFCD00002863; RTECS: OZ4725000 Liquid; Linear Formula: CH2=CH(CH3)CO2CH2CH2OH ; Molecular Formula: C6H10O30; MW: 130.14 Boiling Point: 67°/3.5 mmHg; Flash point: 96°C (205°F) Density (g/mL): 1.073; Refractive Index: 1.453 Air Sensitive


  • RTECS: OZ4725000
  • Refractive index: 1.453
  • Air Sensitive
  • Liquid

Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 67 °C/3.5 mmHg
Density: 1.073 g/mL
Flash Pt: 96 °C (205 °F)
MDL Number: MFCD00002863
CAS Number: 868-77-9
EINECS: 212-782-2

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR