(4-Aminophenyl)acetic acid ethyl ester ≥98%
Supplier: BeanTown Chemical
211870-25G
211870-5G
BT211870-25GEA
383
USD
BT211870-25G
BT211870-5G
(4-Aminophenyl)acetic acid ethyl ester ≥98%
(4-Aminophenyl)acetic acid ethyl ester
CAS: 5438-70-0; MDL No: MFCD00017569
Powder; Molecular Formula: C10H13NO2; MW: 179.22
Melting Point: 46-48°
- Powder
Formula:
C₁₀H₁₃NO₂ Melting Pt: 46…48 °C |
MDL Number:
MFCD00017569 CAS Number: 5438-70-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...